Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Specificity: ABS 061-09-02 show less than 5% cross-Specificity with oxyntomodulin.
Reactivity: ABS 061-09-02 binds free glucagon in solution. ABS 061-09-02 can be used as a detection antibody in a sandwich ELISA with capture antibody ABS 058-08-02.
NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
Glucagon is a 29-residue polypeptide hormone (MW 3482), produced in the pancreas. A related hormone, enteroglucagon (or oxyntomodulin), which is produced in the mucosa of the small and large intestine, consists of the 29 amino acid sequence of pancreatic glucagon extended by 8 additional residues at the C-terminus. The biological activities of pancreatic glucagon include glycogenolysis, lipolysis, gluconeogenesis, and ketogensis, which are antagonistic effects to those of insulin action, thus leading to increased blood glucose levels. Immunocytochemical studies have revealed the presence of pancreatic glucagon inside the A or alpha cells, which constitute 15-20% of the islet cell population. These cells are located preferentially at the periphery of the human pancreatic islets. Pathological manifestations of the glucagon-type peptide residue almost exclusively with the exsistence of tumors or glucagonomas, as no states of glucagon-cell deficiency or hyperplasia have been identified. Glucagon-specific antibodies would prove useful as a cell and tumor markers applying immunohistochemical techniques, and as an analytical tool in qualification of the hormone.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glicentin-related polypeptide; GLP1/2; glucagon; glucagon-like peptide 1; glucagon-like peptide 2; glucagon-like peptide-1; IL12p35; IL12P40; OXY; porcine proglucagon; preproglucagon; Pro-glucagon; proglucagon
Gene Aliases: GCG; GLP-1; GLP1; GLP2; GRPP
UniProt ID: (Human) P01275, (Pig) P01274
Entrez Gene ID: (Human) 2641, (Pig) 397595, (Rat) 24952
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support